Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Difluorophenyl Isocyanate 98.0+%, TCI America™
SDP

Catalog No. D36961G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36961G 1 g
D36965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D36961G Supplier TCI America Supplier No. D36961G

Chemical Identifiers

CAS 42601-04-7
Molecular Formula C7H3F2NO
Molecular Weight (g/mol) 155.104
MDL Number MFCD00065711
InChI Key YBWJVFMNWMLZEN-UHFFFAOYSA-N
Synonym 3,4-difluorophenyl isocyanate, 3,4-difluorophenylisocyanate, isocyanic acid 3,4-difluorophenyl ester, 3,4-difluoro phenyl isocyanate, benzene,1,2-difluoro-4-isocyanato, 1,2-difluoro-4-isocyanato-benzene, 3,4-difluorobenzenisocyanate, acmc-1apvi, 3,4-difluoro-phenylisocyanate, 3,4-difluoro-phenyl isocyanate
PubChem CID 2734708
IUPAC Name 1,2-difluoro-4-isocyanatobenzene
SMILES C1=CC(=C(C=C1N=C=O)F)F

Specifications

Color Yellow
Boiling Point 85°C
Quantity 1 g
UN Number 2206
Formula Weight 155.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,4-Difluorophenyl Isocyanate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.