Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Difluorophenylmagnesium bromide, Thermo Scientific Chemicals

Catalog No. AC399511000
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC399511000 100 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC399511000 Supplier Thermo Scientific Chemicals Supplier No. OMIDE

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90897-92-0 , 109-99-9
Molecular Formula C6H3BrF2Mg
Molecular Weight (g/mol) 217.30
MDL Number MFCD00672004
InChI Key WZHAVYGMKGPNFN-UHFFFAOYSA-M
Synonym 3,4-difluorophenylmagnesium bromide, bromo 3,4-difluorophenyl magnesium, 3,4-difluorophenyl magnesium bromide, c6h3brf2m, wzhavygmkgpnfn-uhfffaoysa-m, magnesium,bromo 3,4-difluorophenyl, 3,4-difluorophenylmagnesium bromide, 0.50 m in 2-methf, 3,4-difluorophenylmagnesium bromide 0.5 m in tetrahydrofuran, 3,4-difluorophenylmagnesium bromide solution, 0.5 m in thf, grignard reagent
PubChem CID 2778383
IUPAC Name magnesium;1,2-difluorobenzene-5-ide;bromide
SMILES FC1=CC=C([Mg]Br)C=C1F

Specifications

CAS Min % 9.01, 86.49
CAS Max % 13.51, 90.99
Color Brown to Yellow
Density 96%5g/mL
Packaging AcroSeal™ Glass Bottle
Linear Formula F2C6H3MgBr
Quantity 100 mL
Formula Weight 217.29
Physical Form Solution
Chemical Name or Material 3, 4-Difluorophenylmagnesium bromide, 0.5M solution in THF
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
May form explosive peroxides.
Reacts violently with water.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Store in a dry place. Store in a closed container.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.