Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dihydro-2H-1,5-benzodioxepine-7-carbaldehyde, 95+%, Thermo Scientific™

Catalog No. p-4510111
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC13204DA 1 g Amber Glass Bottle
CC13204DE 5 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC13204DA Supplier Thermo Scientific Chemicals Supplier No. CC13204DA
Only null left
Add to Cart

Chemical Identifiers

CAS 67869-90-3
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.187
InChI Key LCSVYSVGXQQHSI-UHFFFAOYSA-N
PubChem CID 2776388
IUPAC Name 3,4-dihydro-2H-1,5-benzodioxepine-7-carbaldehyde
SMILES C1COC2=C(C=C(C=C2)C=O)OC1

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 2h-1,5-benzodioxepin-7-carboxaldehyde, 3,4-dihydro, 3,4-dihydro-2h-benzo b 1,4 dioxepine-7-carbaldehyde, 3,4-dihydro-2h-1,5-benzodioxepin-7-carboxaldehyde, 2h-1,5-benzodioxepin-7-carboxaldehyde,3,4-dihydro, 2h,3h,4h-benzo b 1,4-dioxepane-7-carbaldehyde, 3,4-dihydro-2h-1,5-benzodioxepine-7-carboxaldehyde, 3,4-dihydro-2h-benzo b 1,4 dioxepine-7-carbaldehyd
Formula Weight 178.19
Percent Purity 95%
Chemical Name or Material 3, 4-Dihydro-2H-1, 5-benzodioxepine-7-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.