Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Diisopropoxy-3-cyclobutene-1,2-dione 98.0+%, TCI America™
SDP

Catalog No. D22075G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22075G 5 g
D220725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D22075G Supplier TCI America Supplier No. D22075G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 61699-62-5
Molecular Formula C10H14O4
Molecular Weight (g/mol) 198.22
MDL Number MFCD00075125
InChI Key KCZPGGVPQXQGEJ-UHFFFAOYSA-N
Synonym Diisopropyl Squarate
PubChem CID 182314
IUPAC Name bis(propan-2-yloxy)cyclobut-3-ene-1,2-dione
SMILES CC(C)OC1=C(OC(C)C)C(=O)C1=O

Specifications

Melting Point 46°C
Color White
Quantity 5 g
Formula Weight 198.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Diisopropoxy-3-cyclobutene-1,2-dione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.