Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dimethoxybenzenethiol 93.0+%, TCI America™
SDP

Catalog No. D34695G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D34695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D34695G Supplier TCI America Supplier No. D34695G

Chemical Identifiers

CAS 700-96-9
Molecular Formula C8H10O2S
Molecular Weight (g/mol) 170.226
MDL Number MFCD00052823
InChI Key MTKAJLNGIVXZIS-UHFFFAOYSA-N
Synonym 3,4-dimethoxythiophenol, 3,4-dimethoxy thiophenol, 3,4-dimethoxybenzene-1-thiol, 3,4-dimethoxy-thiophenol, benzenethiol, 3,4-dimethoxy, 4-mercaptoveratrole, pubchem6823, acmc-1bdpf, 3,4-dimethoxy-benzenethiol, 3,4-dimethoxyphenylmercaptan
PubChem CID 136539
IUPAC Name 3,4-dimethoxybenzenethiol
SMILES COC1=C(C=C(C=C1)S)OC

Specifications

Color Yellow
Boiling Point 138°C
Quantity 5 g
Formula Weight 170.23
Percent Purity ≥93.0% (T)
Physical Form Liquid
Chemical Name or Material 3,4-Dimethoxybenzenethiol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.