accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 700-96-9

IUPAC Name: 3,4-dimethoxybenzene-1-thiol
Synonyms: 3,4-dimethoxybenzenethiol
Molecular Weight (g/mol): 170.23
Molecular Formula: C8H10O2S
InChi Key: MTKAJLNGIVXZIS-UHFFFAOYSA-N
SMILES: COC1=CC=C(S)C=C1OC
Percent Purity 
  • (1)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)