Learn More
Medchemexpress LLC HY-N1778 100mg Medchemexpress, 3,4-Dimethoxycinnamic acid CAS:2316-26-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN1778100MG
Encompass_Preferred
Medchemexpress, HY-N1778 100mg 3,4-Dimethoxycinnamic acid CAS:2316-26-9 3,4-Dimethoxycinnamic acid (O-Methylferulic acid) is a monomer extracted and purified from Securidaca inappendiculata Hassk. 3,4-Dimethoxycinnamic acid exerts anti-apoptotic effects on L-02 cells via the ROS-mediated signaling pathway [1] . Anti-apoptotic effects [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3854
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.