Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3',4'-Dimethoxyflavone 98.0+%, TCI America™
SDP

Catalog No. D47941G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47941G 1 g
D4794200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D47941G Supplier TCI America Supplier No. D47941G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4143-62-8
Molecular Formula C17H14O4
Molecular Weight (g/mol) 282.295
MDL Number MFCD00143009
InChI Key ZGHORMOOTZTQFL-UHFFFAOYSA-N
Synonym 3',4'-dimethoxyflavone, 2-3,4-dimethoxyphenyl-4h-chromen-4-one, 2-3,4-dimethoxyphenyl chromen-4-one, chembl91153, 4h-1-benzopyran-4-one, 2-3,4-dimethoxyphenyl, 3',4'-dmf, pubchem9847, spectrum_001039, acmc-1apwv, specplus_000555
PubChem CID 688674
IUPAC Name 2-(3,4-dimethoxyphenyl)chromen-4-one
SMILES COC1=C(C=C(C=C1)C2=CC(=O)C3=CC=CC=C3O2)OC

Specifications

Melting Point 153°C
Color White-Yellow
Quantity 1 g
Formula Weight 282.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3′,4′-Dimethoxyflavone
RTECSNumber DJ3012950
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.