Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dimethoxyphenol 97.0+%, TCI America™
SDP

Catalog No. D322125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D322125G 25 g
D32215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D322125G Supplier TCI America Supplier No. D322125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2033-89-8
Molecular Formula C8H10O3
Molecular Weight (g/mol) 154.165
InChI Key SMFFZOQLHYIRDA-UHFFFAOYSA-N
Synonym phenol, 3,4-dimethoxy, 4-hydroxyveratrole, 3,4-bis methyloxy phenol, unii-38b43wcu83, 3,4-dimethoxy phenol, 3,4-dimethoxyphel, 3,4,dimethoxyphenol, phenol,4-dimethoxy, 3,4-dimethoxy-pheno, pubchem4113
PubChem CID 16251
IUPAC Name 3,4-dimethoxyphenol
SMILES COC1=C(C=C(C=C1)O)OC

Specifications

Melting Point 80°C
Boiling Point 167°C
Quantity 25 g
Formula Weight 154.17
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Dimethoxyphenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.