Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dimethoxythiophenol, 98%, Thermo Scientific™

Catalog No. AAA1474514
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1474514 25 g
AAA1474503 1 g
AAA1474506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1474514 Supplier Thermo Scientific Chemicals Supplier No. A1474514

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 700-96-9
Molecular Formula C8H10O2S
Molecular Weight (g/mol) 170.226
MDL Number MFCD00052823
InChI Key MTKAJLNGIVXZIS-UHFFFAOYSA-N
Synonym 3,4-dimethoxythiophenol, 3,4-dimethoxy thiophenol, 3,4-dimethoxybenzene-1-thiol, 3,4-dimethoxy-thiophenol, benzenethiol, 3,4-dimethoxy, 4-mercaptoveratrole, pubchem6823, acmc-1bdpf, 3,4-dimethoxy-benzenethiol, 3,4-dimethoxyphenylmercaptan
PubChem CID 136539
IUPAC Name 3,4-dimethoxybenzenethiol
SMILES COC1=C(C=C(C=C1)S)OC

Specifications

Density 1.19
Boiling Point 263°C
Flash Point >113°C (235°F)
Odor Stench
Refractive Index 1.59
Quantity 25 g
Beilstein 1945136
Sensitivity Air Sensitive
Formula Weight 170.23
Percent Purity 98%
Chemical Name or Material 3,4-Dimethoxythiophenol
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.