Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dimethoxytoluene 99.0+%, TCI America™
SDP

Catalog No. D1986100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1986100G 100 g
D198625G 25 g
D1986500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D1986100G Supplier TCI America Supplier No. D1986100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 494-99-5
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.193
MDL Number MFCD00016651
InChI Key GYPMBQZAVBFUIZ-UHFFFAOYSA-N
Synonym 3,4-dimethoxytoluene, homoveratrole, 4-methylveratrole, benzene, 1,2-dimethoxy-4-methyl, 4-methylveratrol, 4-methyl-1,2-dimethoxybenzene, 1,2-dimethoxy-4-methyl-benzene, unii-349x0g2ssf, homoveratrol
PubChem CID 68126
IUPAC Name 1,2-dimethoxy-4-methylbenzene
SMILES CC1=CC(=C(C=C1)OC)OC

Specifications

Color White-Yellow
Quantity 100 g
Formula Weight 152.19
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 3,4-Dimethoxytoluene
EINECSNumber (3)-0610
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.