Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dimethyl-1,2-cyclopentanedione 98.0+%, TCI America™
SDP

Catalog No. D37825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37825G 5 g
D37821G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D37825G Supplier TCI America Supplier No. D37825G

Chemical Identifiers

CAS 13494-06-9
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.155
MDL Number MFCD00143063
InChI Key WGAVDEVFJDQIMZ-UHFFFAOYSA-N
Synonym 3,4-dimethyl-1,2-cyclopentanedione, 3,4-dimethyl-1,2-cyclopentadione, 1,2-cyclopentanedione, 3,4-dimethyl, fema no. 3268, 1,2-cyclopentanedione,3,4-dimethyl, caramel pentadione, 3,4-dimethyl 1,2-cyclopentandione, acmc-209bxe, 3,4-dimethyl-1,2-cyclopentadione, fg
PubChem CID 61633
IUPAC Name 3,4-dimethylcyclopentane-1,2-dione
SMILES CC1CC(=O)C(=O)C1C

Specifications

Melting Point 69°C
Quantity 5 g
Formula Weight 126.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Dimethyl-1,2-cyclopentanedione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.