Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dimethylbenzyl alcohol, 97%

Catalog No. AAB2492606
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2492606 5 g
AAB2492603 1 g
AAB2492614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2492606 Supplier Thermo Scientific Chemicals Supplier No. B2492606
Add to Cart

CAS: 6966-10-5 | C9H12O | 136.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6966-10-5
MDL Number MFCD00004647

Specifications

Melting Point 62°C to 65°C
Boiling Point 218°C to 221°C
Molecular Formula C9H12O
Quantity 5 g
Synonym 3,4-dimethylbenzyl alcohol, 3,4-dimethylphenyl methanol, benzenemethanol, 3,4-dimethyl, 3,4-dimethylphenyl methan-1-ol, pubchem2161, 3,4-dimethylbenzylalcohol, acmc-1b6qp, 3,4-dimethylbenzyl alcoho, 3,4 dimethylbenzyl alcohol, 4-hydroxymethyl-o-xylene
InChI Key OKGZCXPDJKKZAP-UHFFFAOYSA-N
SMILES CC1=C(C=C(C=C1)CO)C
IUPAC Name (3,4-dimethylphenyl)methanol
Molecular Weight (g/mol) 136.19
PubChem CID 23407
Formula Weight 136.19
Percent Purity 97%
Chemical Name or Material 3,4-Dimethylbenzyl alcohol
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 230-175-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.