Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dimethylthiophenol, 98%, Thermo Scientific™

Catalog No. ac307350010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC307350010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC307350010 Supplier Thermo Scientific Chemicals Supplier No. 307350010
Only null left
Add to Cart

Chemical Identifiers

CAS 18800-53-8
Molecular Formula C8H10S
Molecular Weight (g/mol) 138.23
MDL Number MFCD00010023
InChI Key IDKCKPBAFOIONK-UHFFFAOYSA-N
Synonym 3,4-dimethylthiophenol, benzenethiol, 3,4-dimethyl, 3,4-dimethyl thiophenol, o-xylene-4-thiol, 4-mercapto-o-xylene, 3,4-dimethylbenzene-1-thiol, pubchem6812, 3,4-dimethyl-thiophenol, acmc-1bu1r, 3,4-dimethyl-benzenethiol
PubChem CID 87799
IUPAC Name 3,4-dimethylbenzenethiol
SMILES CC1=C(C=C(C=C1)S)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0270g/mL
Boiling Point 218°C
Flash Point 89°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5730 to 1.5750 (20°C, 589nm)
Linear Formula CH3C6H3SHCH3
Quantity 1 g
Specific Gravity 1.027
Formula Weight 138.23
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3, 4-Dimethylthiophenol, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Co
EINECSNumber 242-587-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.