Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Ethylenedioxythiophene-2-carboxaldehyde 98.0+%, TCI America™
SDP

Catalog No. E11501G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11501G 1 g
E11505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E11501G Supplier TCI America Supplier No. E11501G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 204905-77-1
Molecular Formula C7H6O3S
Molecular Weight (g/mol) 170.182
MDL Number MFCD01651766
InChI Key GNVXYRDVJKJZTO-UHFFFAOYSA-N
Synonym 2,3-dihydrothieno 3,4-b 1,4 dioxine-5-carbaldehyde, 2h,3h-thieno 3,4-b 1,4 dioxine-5-carbaldehyde, 3,4-ethylenedioxythiophene-2-carbaldehyde, 3,4-ethylenedioxythiophene-2-carboxaldehyde, 2-formyl-3,4-ethylene dioxythiophene, 2h,3h-thiopheno 3,4-e 1,4-dioxane-5-carbaldehyde, 2,3-dihydrothieno 3,4-b-1,4-dioxin-5-carbaldehyde, 2,3-dihydrothieno 3,4-b 1,4 dioxine-5-carboxaldehyde, thieno 3,4-b-1,4-dioxin-5-carboxaldehyde, 2,3-dihydro
PubChem CID 3540090
IUPAC Name 2,3-dihydrothieno[3,4-b][1,4]dioxine-5-carbaldehyde
SMILES C1COC2=C(SC=C2O1)C=O

Specifications

Melting Point 141°C
Color White-Yellow
Quantity 1 g
Formula Weight 170.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Ethylenedioxythiophene-2-carboxaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.