Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(4-Fluorophenoxy)benzyl Bromide 97.0+%, TCI America™
SDP

Catalog No. F02431G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F02431G 1 g
F02435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F02431G Supplier TCI America Supplier No. F02431G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 65295-58-1
Molecular Formula C13H10BrFO
Molecular Weight (g/mol) 281.124
MDL Number MFCD00059915
InChI Key JGFSTQUUDSBQCO-UHFFFAOYSA-N
PubChem CID 2737456
IUPAC Name 1-(bromomethyl)-3-(4-fluorophenoxy)benzene
SMILES C1=CC(=CC(=C1)OC2=CC=C(C=C2)F)CBr

Specifications

Color White-Yellow
Boiling Point 160°C
Quantity 1 g
Synonym 3′C-Bromomethyl-4-fluorodiphenyl Ether
Formula Weight 281.12
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 3-(4-Fluorophenoxy)benzyl Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.