Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(4-hydroxyphenyl)propanohydrazide, Thermo Scientific™

Catalog No. BTB00937DE
Click to view available options
Quantity:
5 g
10 g

Catalog No. BTB00937DE

Supplier: Thermo Scientific Chemicals BTB00937DE

Only null left
Add to Cart

Chemical Identifiers

CAS 65330-63-4
Molecular Formula C9H12N2O2
Molecular Weight (g/mol) 180.21
MDL Number MFCD00025134
InChI Key JMNHUOPRGLTNJI-UHFFFAOYSA-N
Synonym 3-4-hydroxyphenyl propanohydrazide, 3-4-hydroxyphenyl propanehydrazide, 3-4-hydroxyphenyl propionohydrazide, benzenepropanoic acid,4-hydroxy-, hydrazide, benzenepropanoic acid, 4-hydroxy-, hydrazide
PubChem CID 2796587
IUPAC Name 3-(4-hydroxyphenyl)propanehydrazide
SMILES NNC(=O)CCC1=CC=C(O)C=C1

Specifications

Packaging Amber glass bottle
Quantity 5 g
Formula Weight 180.21
Percent Purity 97%
Chemical Name or Material 3-(4-hydroxyphenyl)propanohydrazide
Hazard Category GHS Signal Word: Warning
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.