Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(4-Methoxybenzoyl)propionic Acid 98.0+%, TCI America™
SDP

Catalog No. M138325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M138325G 25 g
M13835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M138325G Supplier TCI America Supplier No. M138325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3153-44-4
Molecular Formula C11H12O4
Molecular Weight (g/mol) 208.213
MDL Number MFCD00002795
InChI Key OMTDIBZSUZNVJK-UHFFFAOYSA-N
Synonym 3-4-methoxybenzoyl propionic acid, 4-4-methoxyphenyl-4-oxobutanoic acid, 4-4-methoxyphenyl-4-oxobutyric acid, 3-4-methoxybenzoyl propanoic acid, beta-p-methoxybenzoyl propionic acid, 4-4-methoxyphenyl-4-oxo-butyric acid, 4-4-methoxyphenyl-4-oxo-butanoic acid, benzenebutanoic acid, 4-methoxy-gamma-oxo, maybridge1_002003, acmc-1cl1u
PubChem CID 76613
IUPAC Name 4-(4-methoxyphenyl)-4-oxobutanoic acid
SMILES COC1=CC=C(C=C1)C(=O)CCC(=O)O

Specifications

Melting Point 149°C
Color White-Yellow
Quantity 25 g
Formula Weight 208.21
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(4-Methoxybenzoyl)propionic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.