Learn More
Sigma Organic Chemistry 3-4-Methoxyphenyl propa | 100mg | MFCD00463001
Supplier: Sigma Organic Chemistry CDS004132100MG

3-4-Methoxyphenyl propa , 100mg
About this Item:
MDL #: MFCD00463001
Molecular Weight: 194.23
UNSPSC Code: 12352200
Chemical Formula: C10H14N2O2
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.