Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3',4'-(Methylenedioxy)acetophenone, 98%

Catalog No. AAA1359730
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1359730 250 g
AAA1359709 10 g
AAA1359718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1359730 Supplier Thermo Scientific Chemicals Supplier No. A1359730
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 3162-29-6 | C9H8O3 | 164.16 g/mol

3?,4?-(Methylenedioxy)acetophenone is used in the synthesis of N-ethyl-1-(3,4-methylenedioxyphenyl)ethylamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3′,4′-(Methylenedioxy)acetophenone is used in the synthesis of N-ethyl-1-(3,4-methylenedioxyphenyl)ethylamine.

Solubility
Slightly soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3162-29-6
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00005831
InChI Key BMHMKWXYXFBWMI-UHFFFAOYSA-N
Synonym 3',4'-methylenedioxy acetophenone, 1-benzo d 1,3 dioxol-5-yl ethanone, 5-acetyl-1,3-benzodioxole, 3,4-methylenedioxyacetophenone, 3,4-methylenedioxy acetophenone, 3',4'-methylenedioxyacetophenone, 1-1,3-benzodioxol-5-yl ethanone, acetopiperone, 1-1,3-benzodioxol-5-yl ethan-1-one, ethanone, 1-1,3-benzodioxol-5-yl
PubChem CID 76622
IUPAC Name 1-(1,3-benzodioxol-5-yl)ethanone
SMILES CC(=O)C1=CC2=C(C=C1)OCO2

Specifications

Melting Point 85°C to 89°C
Boiling Point 152°C to 155°C (15 mmHg)
Quantity 250 g
Beilstein 139669
Solubility Information Slightly soluble in water.
Formula Weight 164.16
Percent Purity 98%
Chemical Name or Material 3',4'-(Methylenedioxy)acetophenone
EINECSNumber 221-613-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.