Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Methylenedioxy-beta-nitrostyrene, 98%, Thermo Scientific™

Catalog No. AAL0913909
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0913909 10 g
AAL0913918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0913909 Supplier Thermo Scientific Chemicals Supplier No. L0913909

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1485-00-3
Molecular Formula C9H7NO4
Molecular Weight (g/mol) 193.158
MDL Number MFCD00014575
InChI Key KFLWBZPSJQPRDD-ONEGZZNKSA-N
Synonym 3,4-methylenedioxy-beta-nitrostyrene, mns, syk inhibitor iii, 1,3-benzodioxole, 5-nitrovinyl, 5-2-nitrovinyl benzo d 1,3 dioxole, 1,3-benzodioxole, 5-2-nitroethenyl, 5-2-nitrovinyl-1,3-benzodioxole, 5-2-nitroethenyl-1,3-benzodioxole, styrene, 3,4-methylenedioxy-beta-nitro, e-5-2-nitrovinyl benzo d 1,3 dioxole
PubChem CID 672296
IUPAC Name 5-[(E)-2-nitroethenyl]-1,3-benzodioxole
SMILES C1OC2=C(O1)C=C(C=C2)C=C[N+](=O)[O-]

Specifications

Melting Point 159°C to 163°C
Quantity 10 g
Beilstein 192350
Sensitivity Light sensitive
Formula Weight 193.16
Percent Purity 98%
Chemical Name or Material 3,4-Methylenedioxy-beta-nitrostyrene
Hazard Category H302-H315-H319-H335-H500
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
RTECSNumber WL5270000
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.