Learn More
Sigma Organic Chemistry 3 4-Methylenedioxy prop | 250mg | 28281-49-4 | MFCD00016652
Supplier: Sigma Organic Chemistry CDS001550250MG

3 4-Methylenedioxy prop, 250mg
About this Item:
CAS #: 28281-49-4
MDL #: MFCD00016652
Molecular Weight: 178.18
UNSPSC Code: 12352200
Chemical Formula: C10H10O3
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.