Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4'-Oxydiphthalic Anhydride 98.0+%, TCI America™
SDP

Catalog No. O03841G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O03841G 1 g
O038425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O03841G Supplier TCI America Supplier No. O03841G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 50662-95-8
Molecular Formula C16H6O7
Molecular Weight (g/mol) 310.22
MDL Number MFCD22460472
InChI Key OPVHOFITDJSMOD-UHFFFAOYSA-N
PubChem CID 14574292
IUPAC Name 4-[(1,3-dioxo-1,3-dihydro-2-benzofuran-5-yl)oxy]-1,3-dihydro-2-benzofuran-1,3-dione
SMILES O=C1OC(=O)C2=C1C=CC(OC1=CC=CC3=C1C(=O)OC3=O)=C2

Specifications

Melting Point 180°C
Color White-Yellow
Quantity 1 g
Formula Weight 310.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,4′-Oxydiphthalic Anhydride
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.