Learn More
Sigma Organic Chemistry 3- 4-Pyridinyl propanoi | 50mg | 6318-43-0 | MFCD00084846
Supplier: Sigma Organic Chemistry CDS00209450MG

3- 4-Pyridinyl propanoi , 50mg
About this Item:
CAS #: 6318-43-0
MDL #: MFCD00084846
Molecular Weight: 151.16
UNSPSC Code: 12352200
Chemical Formula: C8H9NO2
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.