Learn More
Alkali Scientific ASI™ 3/4" wide x 500" Color Coding Labeling Tape, 500 per Roll, 16/Rolls, Dark Green
SDP
Supplier:  Alkali Scientific TC75DARK GREEN
Encompass_Preferred
Color Coding Labels is our another popular item, which is offered in an assortment of colors. This tape is beneficial in any laboratory, medical, or educational setting. From labeling test tubes to laboratory wraps, our brand color coding tape is an essential! Strong and Durable – oil, acid, and water resistant Withstands temperatures from -23º C to +121ºC (-10º F to +250ºF) Can be written on with pen, pencil, or permanent marker without smearing Available in 27 different colors and 1/2”, 3/4”, 1”, 1 1/2”, and 2” widths Packing: 16 Rolls/case Labels Per Case: 500"/Roll, 16 Rolls/case
Catalog No. 50-302-8778
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.