Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5,6-Trichlorosalicylic Acid 97.0+%, TCI America™
SDP

Catalog No. T140625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T140625G 25 g
T1406500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T140625G Supplier TCI America Supplier No. T140625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 40932-60-3
Molecular Formula C7H3Cl3O3
Molecular Weight (g/mol) 241.448
MDL Number MFCD00075245
InChI Key IIHCUZVBIMTHEB-UHFFFAOYSA-N
Synonym 3,5,6-trichlorosalicylic acid, benzoic acid, 2,3,5-trichloro-6-hydroxy, 3,5,6-trichloro-2-hydroxybenzoic acid, ghl.pd_mitscher_leg0.102, acmc-209jhp, ksc489o8r, chembl10381, 3,5,6-trichloro-salicylic acid, 2,3,5-trichloro-6-hydroxy-benzoic acid, #
PubChem CID 94511
IUPAC Name 2,3,5-trichloro-6-hydroxybenzoic acid
SMILES C1=C(C(=C(C(=C1Cl)Cl)C(=O)O)O)Cl

Specifications

Melting Point 208°C
Quantity 25 g
Formula Weight 241.45
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5,6-Trichlorosalicylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.