Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Bis(trifluoromethyl)benzyl alcohol, 98%

Catalog No. AAA1929006
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1929006 5 g
AAA1929003 1 g
AAA1929014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1929006 Supplier Thermo Scientific Chemicals Supplier No. A1929006
Add to Cart

CAS: 32707-89-4 | C9H6F6O | 244.136 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32707-89-4
Molecular Formula C9H6F6O
Molecular Weight (g/mol) 244.136
MDL Number MFCD00009907
InChI Key BJTWPJOGDWRYDD-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl benzyl alcohol, 3,5-bis trifluoromethyl phenyl methanol, 3,5-bis-trifluoromethyl-phenyl-methanol, 3,5-ditrifluoromethylbenzyl alcohol, benzenemethanol, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl benzylalcohol, 3,5-di trifluoromethyl benzyl alcohol, 3,5-di trifluoromethyl phenyl methanol, 3,5-bis-trifluoromethyl benzyl alcohol
PubChem CID 122933
IUPAC Name [3,5-bis(trifluoromethyl)phenyl]methanol
SMILES C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)CO

Specifications

Melting Point 52°C to 56°C
Boiling Point 40°C (0.5 mmHg)
Flash Point 97°C (206°F)
Quantity 5 g
Beilstein 2055358
Formula Weight 244.15
Percent Purity 98%
Chemical Name or Material 3,5-Bis(trifluoromethyl)benzyl alcohol
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 251-168-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.