Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Bis(trifluoromethyl)phenol 96.0+%, TCI America™
SDP

Catalog No. B186925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B186925G 25 g
B18695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B186925G Supplier TCI America Supplier No. B186925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 349-58-6
Molecular Formula C8H4F6O
Molecular Weight (g/mol) 230.109
MDL Number MFCD00000386
InChI Key ODSXJQYJADZFJX-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl phenol, phenol, 3,5-bis trifluoromethyl, 3,5-di trifluoromethyl phenol, 3,5-bis-trifluoromethylphenol, 3,5-ditrifluoromethylphenol, 3,5-bis-trifluoromethyl phenol, alpha,alpha,alpha,alpha',alpha',alpha'-hexafluoro-3,5-xylenol, pubchem12483, mbt-oh, acmc-1clsr
PubChem CID 67680
IUPAC Name 3,5-bis(trifluoromethyl)phenol
SMILES C1=C(C=C(C=C1C(F)(F)F)O)C(F)(F)F

Specifications

Color Yellow
Boiling Point 88°C
Quantity 25 g
Formula Weight 230.11
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,5-Bis(trifluoromethyl)phenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.