Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Di(1-pyrenyl)pyridine 98.0+%, TCI America™
SDP

Catalog No. D49311G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49311G 1 g
D4931200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D49311G Supplier TCI America Supplier No. D49311G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1246467-58-2
Molecular Formula C37H21N
Molecular Weight (g/mol) 479.582
InChI Key YNTQHSDEZACXAP-UHFFFAOYSA-N
PubChem CID 102472598
IUPAC Name 3,5-di(pyren-1-yl)pyridine
SMILES C1=CC2=C3C(=C1)C=CC4=C(C=CC(=C43)C=C2)C5=CC(=CN=C5)C6=C7C=CC8=CC=CC9=C8C7=C(C=C9)C=C6

Specifications

Color Yellow
Quantity 1 g
Formula Weight 479.58
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Di(1-pyrenyl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.