Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Di-tert-butyl-4-hydroxybenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog No. AC161211000
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC161211000 100 g
AC161210250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC161211000 Supplier Thermo Scientific Chemicals Supplier No. 161211000

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1620-98-0
MDL Number MFCD00008826
InChI Key DOZRDZLFLOODMB-UHFFFAOYSA-N
Synonym 3,5-di-tert-butyl-4-hydroxybenzaldehyde, 3,5-di-t-butyl-4-hydroxybenzaldehyde, unii-95vti93vul, benzaldehyde, 3,5-bis 1,1-dimethylethyl-4-hydroxy, benzaldehyde, 3,5-di-tert-butyl-4-hydroxy, 95vti93vul, 3,5-ditert-butyl-4-hydroxy-benzaldehyde, 3,5-di tert-butyl-4-hydroxybenzaldehyde, 3,5-di-tert-butyl-4-hydroxy-benzaldehyde, 3,5-bis tert-butyl-4-hydroxybenzaldehyde
PubChem CID 73219
IUPAC Name 3,5-ditert-butyl-4-hydroxybenzaldehyde
SMILES CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)C=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 186°C to 190°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C15H22O2
Quantity 100 g
Molecular Weight (g/mol) 234.339
Formula Weight 234.34
Percent Purity ≥96.0%
Physical Form Crystalline Powder
Chemical Name or Material 3, 5-Di-tert-butyl-4-hydroxybenzaldehyde, 97%
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 216-592-0
RTECSNumber CU5610070
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.