Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Di-tert-butyltoluene 98.0+%, TCI America™
SDP

Catalog No. D341425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D341425G 25 g
D34145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D341425G Supplier TCI America Supplier No. D341425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15181-11-0
Molecular Formula C15H24
Molecular Weight (g/mol) 204.357
MDL Number MFCD00026300
InChI Key WIXDSJRJFDWTNY-UHFFFAOYSA-N
Synonym 3,5-di-tert-butyltoluene, 1,3-di-tert-butyl-5-methylbenzene, toluene, 3,5-di-tert-butyl, 3,5-bis tert-butyl toluene, benzene, 1,3-bis 1,1-dimethylethyl-5-methyl, 3,5-di-t-butyltoluene, 3,5-bis t-butyl-1-methylbenzene, 3,5-di-tert-butyl-1-methylbenzene, 1,3-bis tert-butyl-5-methylbenzene
PubChem CID 84819
IUPAC Name 1,3-ditert-butyl-5-methylbenzene
SMILES CC1=CC(=CC(=C1)C(C)(C)C)C(C)(C)C

Specifications

Color White-Yellow
Boiling Point 244°C
Quantity 25 g
Formula Weight 204.36
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 3,5-Di-tert-butyltoluene
EINECSNumber (3)-3427
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.