Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Diamino-1,2,4-triazole 97.0+%, TCI America™
SDP

Catalog No. D258725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D258725G 25 g
D25875G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D258725G Supplier TCI America Supplier No. D258725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1455-77-2
Molecular Formula C2H5N5
Molecular Weight (g/mol) 99.097
MDL Number MFCD00005233
InChI Key PKWIYNIDEDLDCJ-UHFFFAOYSA-N
Synonym guanazole, 3,5-diamino-1,2,4-triazole, 4h-1,2,4-triazole-3,5-diamine, 3,5-diamino-1h-1,2,4-triazole, 3,5-diamino-s-triazole, s-triazole, 3,5-diamino, 1,2,4-triazolidine, 3,5-diimino, unii-i01twm5267, ccris 3745, 3-amino-1h-1,2,4-triazol-5-ylamine
PubChem CID 15078
ChEBI CHEBI:75425
IUPAC Name 1H-1,2,4-triazole-3,5-diamine
SMILES C1(=NC(=NN1)N)N

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 99.10
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Diamino-1,2,4-triazole
EINECSNumber (5)-3586
RTECSNumber XZ4535000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.