Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dibenzyloxybenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. D226725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D226725G 25 g
D22675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D226725G Supplier TCI America Supplier No. D226725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 28917-43-3
Molecular Formula C21H18O4
Molecular Weight (g/mol) 334.371
MDL Number MFCD00191341
InChI Key DHQIBPUGSWVDOH-UHFFFAOYSA-N
Synonym 3,5-Bis(benzyloxy)benzoic Acid
PubChem CID 99307
IUPAC Name 3,5-bis(phenylmethoxy)benzoic acid
SMILES C1=CC=C(C=C1)COC2=CC(=CC(=C2)C(=O)O)OCC3=CC=CC=C3

Specifications

Melting Point 214°C
Color White-Yellow
Quantity 25 g
Formula Weight 334.37
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dibenzyloxybenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.