Learn More
Medchemexpress LLC 3,5-DiBr-PAESA sodium | 164581-25-3 | 528.19 g/mol | C16H17Br2N4NaO3S | 10 MG
SDP
Supplier:  Medchemexpress LLC HYD1895A10MG
Encompass_Preferred
3,5-DiBr-PAESA sodium is an ultrasensitive chelating agent used for the determination of silver(I) in water and, in the absence of interfering ions, for the determination of copper(II). Supplied as a brown-to-black solid for analytical and biochemical applications, it is offered in small laboratory package sizes for research use.
  • Ultrasensitive chelating agent for silver(I) detection
  • Also applicable for copper(II) determination
  • Solid reagent with brown-to-black appearance
  • Store protected from light; solvent storage guidance provided
  • Available in small laboratory sizes, including 10 mg
  • Molecular formula C16H17Br2N4NaO3S; molecular weight 528.19 g/mol
Catalog No. 50-002-97563