Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dibromo-1,2,4-triazole 97.0+%, TCI America™
SDP

Catalog No. D503025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D503025G 25 g
D50305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D503025G Supplier TCI America Supplier No. D503025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7411-23-6
Molecular Formula C2HBr2N3
Molecular Weight (g/mol) 226.859
MDL Number MFCD00487260
InChI Key FRAKFBWDPXYIQO-UHFFFAOYSA-N
Synonym 3,5-dibromo-4h-1,2,4-triazole, 3,5-dibromo-1,2,4-triazole, 1h-1,2,4-triazole, 3,5-dibromo, s-triazole, 3,5-dibromo, 3,5-dibromo-s-triazole, so, deblqxbskfssp@, pubchem13285, acmc-1bktu, 1h-1,4-triazole, 3,5-dibromo
PubChem CID 81904
IUPAC Name 3,5-dibromo-1H-1,2,4-triazole
SMILES C1(=NC(=NN1)Br)Br

Specifications

Melting Point 212°C
Quantity 25 g
Formula Weight 226.86
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dibromo-1,2,4-triazole
RTECSNumber XZ4570000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.