Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dibromo-2-methylthiophene 95.0+%, TCI America™
SDP

Catalog No. D399925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D399925G 25 g
D39995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D399925G Supplier TCI America Supplier No. D399925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 29421-73-6
Molecular Formula C5H4Br2S
Molecular Weight (g/mol) 255.96
MDL Number MFCD07781197
InChI Key OGAJGUIMFMRGRB-UHFFFAOYSA-N
PubChem CID 122373
IUPAC Name 3,5-dibromo-2-methylthiophene
SMILES CC1=C(Br)C=C(Br)S1

Specifications

Melting Point -15°C
Color Yellow
Boiling Point 230°C
Quantity 25 g
Formula Weight 255.96
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,5-Dibromo-2-methylthiophene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.