Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3',5'-Dibromo-4'-hydroxyacetophenone 97.0+%, TCI America™
SDP

Catalog No. D282425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D282425G 25 g
D28245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D282425G Supplier TCI America Supplier No. D282425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2887-72-1
Molecular Formula C8H6Br2O2
Molecular Weight (g/mol) 293.94
MDL Number MFCD00075779
InChI Key ZNWPTJSBHHIXLJ-UHFFFAOYSA-N
PubChem CID 76158
IUPAC Name 1-(3,5-dibromo-4-hydroxyphenyl)ethan-1-one
SMILES CC(=O)C1=CC(Br)=C(O)C(Br)=C1

Specifications

Color Red-White
Quantity 25 g
Formula Weight 293.94
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3′,5′-Dibromo-4′-hydroxyacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.