Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dibromo-4-iodotoluene, 98%, Thermo Scientific™

Catalog No. p-7080798
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1050203 1 g
AAA1050206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1050203 Supplier Thermo Scientific Chemicals Supplier No. A1050203

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 175278-10-1
Molecular Formula C7H5Br2I
Molecular Weight (g/mol) 375.83
MDL Number MFCD00052397
InChI Key VLVXYJATBIXIPF-UHFFFAOYSA-N
Synonym 3,5-dibromo-4-iodotoluene, buttpark 121\18-24, 2,6-dibromo-4-methyliodobenzene, benzene,1,3-dibromo-2-iodo-5-methyl, pubchem3827, acmc-1c7yh, 1,3-bis bromanyl-2-iodanyl-5-methyl-benzene
PubChem CID 2801325
IUPAC Name 1,3-dibromo-2-iodo-5-methylbenzene
SMILES CC1=CC(Br)=C(I)C(Br)=C1

Specifications

Melting Point 69°C to 72°C
Boiling Point 126°C to 128°C (0.4 mmHg)
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 375.83
Percent Purity 98%
Chemical Name or Material 3,5-Dibromo-4-iodotoluene
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.