Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dibromobenzoic Acid 97.0+%, TCI America™
SDP

Catalog No. D235325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D235325G 25 g
D23535G 5 g
D235310G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D235325G Supplier TCI America Supplier No. D235325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 618-58-6
Molecular Formula C7H4Br2O2
Molecular Weight (g/mol) 279.92
MDL Number MFCD00051758
InChI Key SFTFNJZWZHASAQ-UHFFFAOYSA-N
Synonym benzoic acid, 3,5-dibromo, 3,5-dibromobenzoicacid, benzoicacid, 3,5-dibromo, buttpark 99\57-19, pubchem3861, acmc-209mwz, maybridge1_003318, 3,5-dibromo-benzoic acid, 4-09-00-01028 beilstein handbook reference, ksc358a3h
PubChem CID 12063
IUPAC Name 3,5-dibromobenzoic acid
SMILES OC(=O)C1=CC(Br)=CC(Br)=C1

Specifications

Melting Point 220°C
Color White-Yellow
Quantity 25 g
Formula Weight 279.92
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dibromobenzoic Acid
EINECSNumber (3)-0062
RTECSNumber DG6290010
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.