Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dibromotoluene 98.0+%, TCI America™
SDP

Catalog No. D337625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D337625G 25 g
D33765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D337625G Supplier TCI America Supplier No. D337625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1611-92-3
Molecular Formula C7H6Br2
Molecular Weight (g/mol) 249.93
MDL Number MFCD00013528
InChI Key DPKKOVGCHDUSAI-UHFFFAOYSA-N
Synonym 3,5-dibromotoluene, benzene, 1,3-dibromo-5-methyl, toluene, 3,5-dibromo, 3 ,5-dibromotoluene, 1,3-dibromo-5-methyl-benzene, 5-dibromotoluene, 3,5-dibromo toluene, 3,5-dibromo-toluene, pubchem3934, acmc-1bs6w
PubChem CID 15361
IUPAC Name 1,3-dibromo-5-methylbenzene
SMILES CC1=CC(Br)=CC(Br)=C1

Specifications

Melting Point 41°C
Color White-Yellow
Boiling Point 246°C
Quantity 25 g
Formula Weight 249.93
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dibromotoluene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.