Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dichloro-2-cyanopyridine, 95%, Thermo Scientific Chemicals

Catalog No. AAL1972314
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1972314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1972314 Supplier Thermo Scientific Chemicals Supplier No. L1972314
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 85331-33-5 | Molecular Formula: C6H2Cl2N2 | Molecular Weight: 172.996 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85331-33-5
Molecular Formula C6H2Cl2N2
Molecular Weight (g/mol) 172.996
MDL Number MFCD03788758
InChI Key ATUOLSDAAPMVJJ-UHFFFAOYSA-N
Synonym 3,5-dichloro-2-cyanopyridine, 2-cyano-3,5-dichloropyridine, 3,5-dichloropicolinonitrile, 2-pyridinecarbonitrile, 3,5-dichloro, pubchem2423, acmc-209q5l, ksc448a0l, tpc-py042, 3,5-dichloro-2-cyano pyridine, 3,5-dichloro-2-pyridinecarbonitrile
PubChem CID 2769699
IUPAC Name 3,5-dichloropyridine-2-carbonitrile
SMILES C1=C(C=NC(=C1Cl)C#N)Cl

Specifications

Melting Point 99°C to 103°C
Quantity 25 g
UN Number UN3439
Beilstein 4390101
Formula Weight 173
Percent Purity 95%
Chemical Name or Material 3,5-Dichloro-2-cyanopyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.