Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dichloro-2-cyanopyridine 98.0+%, TCI America™
SDP

Catalog No. D40595G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40595G 5 g
D40591G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D40595G Supplier TCI America Supplier No. D40595G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 85331-33-5
Molecular Formula C6H2Cl2N2
Molecular Weight (g/mol) 172.996
MDL Number MFCD03788758
InChI Key ATUOLSDAAPMVJJ-UHFFFAOYSA-N
Synonym 3,5-dichloro-2-cyanopyridine, 2-cyano-3,5-dichloropyridine, 3,5-dichloropicolinonitrile, 2-pyridinecarbonitrile, 3,5-dichloro, pubchem2423, acmc-209q5l, ksc448a0l, tpc-py042, 3,5-dichloro-2-cyano pyridine, 3,5-dichloro-2-pyridinecarbonitrile
PubChem CID 2769699
IUPAC Name 3,5-dichloropyridine-2-carbonitrile
SMILES C1=C(C=NC(=C1Cl)C#N)Cl

Specifications

Melting Point 102°C
Color Yellow
Quantity 5 g
UN Number 3439
Formula Weight 173.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dichloro-2-cyanopyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.