Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dichloro-2-fluoropyridine 98.0+%, TCI America™
SDP

Catalog No. D33171G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33171G 1 g
D33175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D33171G Supplier TCI America Supplier No. D33171G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 823-56-3
Molecular Formula C5H2Cl2FN
Molecular Weight (g/mol) 165.98
MDL Number MFCD04039349
InChI Key FHQWUIZMJXPGRG-UHFFFAOYSA-N
PubChem CID 2783256
IUPAC Name 3,5-dichloro-2-fluoropyridine
SMILES FC1=NC=C(Cl)C=C1Cl

Specifications

Melting Point 44°C
Color White-Yellow
Quantity 1 g
Synonym 2-fluoro-3,5-dichloropyridine, pyridine, 3,5-dichloro-2-fluoro, 3,5-dichloro-6-fluoropyridine, pubchem6240, acmc-209poe, ksc649k9h, 3,5-dichloro-2-fluoro pyridine, 3,5-dichloro-2-fluoro-pyridine, fhqwuizmjxpgrg-uhfffaoysa, pyridine,3,5-dichloro-2-fluoro
Formula Weight 165.98
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dichloro-2-fluoropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.