Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dichloro-4-iodopyridine 98.0+%, TCI America™
SDP

Catalog No. D51951G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D51951G 1 g
D51955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D51951G Supplier TCI America Supplier No. D51951G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 343781-41-9
Molecular Formula C5H2Cl2IN
Molecular Weight (g/mol) 273.88
MDL Number MFCD03094687
InChI Key DEXLTMBFLWCGNF-UHFFFAOYSA-N
PubChem CID 2736004
IUPAC Name 3,5-dichloro-4-iodopyridine
SMILES ClC1=CN=CC(Cl)=C1I

Specifications

Melting Point 189°C
Quantity 1 g
Formula Weight 273.88
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dichloro-4-iodopyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.