Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dichloroisonicotinic Acid 98.0+%, TCI America™
SDP

Catalog No. D26005G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26005G 5 g
D26001G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D26005G Supplier TCI America Supplier No. D26005G

Chemical Identifiers

CAS 13958-93-5
Molecular Formula C6H3Cl2NO2
Molecular Weight (g/mol) 191.995
MDL Number MFCD01861975
InChI Key BUQPTOSHKHYHHB-UHFFFAOYSA-N
Synonym 3,5-dichloroisonicotinic acid, 3,5-dichloro-4-pyridinecarboxylic acid, 4-pyridinecarboxylic acid, 3,5-dichloro, dichloroisonicotinicacid, acmc-1cbzu, 3?5-dichloroisonicotinic acid, 3,5-dichloro-isonicotinic acid, buqptoshkhyhhb-uhfffaoysa, 3,5-dichloroisonicotinic acid #, 3,5-dichloro-pyridine-4-carboxylic acid
PubChem CID 605763
IUPAC Name 3,5-dichloropyridine-4-carboxylic acid
SMILES C1=C(C(=C(C=N1)Cl)C(=O)O)Cl

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 192.00
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dichloroisonicotinic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.