Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dichlorophenylhydrazine Hydrochloride 90.0+%, TCI America™
SDP

Catalog No. D264425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D264425G 25 g
D26445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D264425G Supplier TCI America Supplier No. D264425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 63352-99-8
Molecular Formula C6H7Cl3N2
Molecular Weight (g/mol) 213.486
MDL Number MFCD00012938
InChI Key BDXZOJVMTJOAPS-UHFFFAOYSA-N
Synonym 3,5-dichlorophenylhydrazine hydrochloride, 3,5-dichlorophenyl hydrazine hydrochloride, 3,5-dichlorophenyl hydrazine monohydrochloride, 3,5-dichlorophenylhydrazine hcl, 1-3,5-dichlorophenyl hydrazine hydrochloride, hydrazine, 3,5-dichlorophenyl-, monohydrochloride, 3,5-dichlorophenylhydrazinehydrochloride, pubchem7531, c6h6cl2n2.hcl, acmc-209nf3
PubChem CID 2723913
IUPAC Name (3,5-dichlorophenyl)hydrazine;hydrochloride
SMILES C1=C(C=C(C=C1Cl)Cl)NN.Cl

Specifications

Quantity 25 g
Formula Weight 213.49
Percent Purity ≥90.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dichlorophenylhydrazine Hydrochloride
EINECSNumber (3)-0472
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.