Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dichloropyridine-2-carboxamide, 97%, Thermo Scientific Chemicals

Catalog No. AAH6698103
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6698103 1 g
AAH66981MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6698103 Supplier Thermo Scientific Chemicals Supplier No. H6698103
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5468-71-3
Molecular Formula C6H4Cl2N2O
Molecular Weight (g/mol) 191.011
MDL Number MFCD20529142
InChI Key QIRSAVRBVNYEMF-UHFFFAOYSA-N
Synonym 3,5-dichloropicolinamide, 3,5-dichloropicolinic acid amide, 3,5-dichloro-pyridine-2-carboxylic acid amide
PubChem CID 225522
IUPAC Name 3,5-dichloropyridine-2-carboxamide
SMILES C1=C(C=NC(=C1Cl)C(=O)N)Cl

Specifications

Color White
Quantity 1 g
Beilstein 136067
Formula Weight 191.01
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3,5-Dichloropyridine-2-carboxamide
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.