Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Difluorobenzoyl Chloride 98.0+%, TCI America™
SDP

Catalog No. D340725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D340725G 25 g
D34075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D340725G Supplier TCI America Supplier No. D340725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 129714-97-2
Molecular Formula C7H3ClF2O
Molecular Weight (g/mol) 176.547
MDL Number MFCD00010309
InChI Key OYZWEOORLJBPMA-UHFFFAOYSA-N
Synonym 3,5-difluorobenzoylchloride, benzoyl chloride, 3,5-difluoro, 3,5-difluorobenzoic acid chloride, pubchem20777, acmc-1chrc, intermediates-zcf02002, 3,5difluorobenzoyl chloride, 3,5-di-fluorobenzoyl chloride, 3,5-difluoro-benzoyl chloride, 3,5-difluoro-benzoyl-chloride
PubChem CID 145600
IUPAC Name 3,5-difluorobenzoyl chloride
SMILES C1=C(C=C(C=C1F)F)C(=O)Cl

Specifications

Color Yellow
Quantity 25 g
UN Number 3265
Formula Weight 176.55
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3,5-Difluorobenzoyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.