Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Difluorobenzylamine 98.0+%, TCI America™
SDP

Catalog No. D33435G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33435G 5 g
D33431G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D33435G Supplier TCI America Supplier No. D33435G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 90390-27-5
Molecular Formula C7H7F2N
Molecular Weight (g/mol) 143.14
MDL Number MFCD00061244
InChI Key VJNGGOMRUHYAMC-UHFFFAOYSA-N
Synonym 3,5-difluorobenzylamine, 3,5-difluorophenyl methanamine, benzenemethanamine, 3,5-difluoro, 1-3,5-difluorophenyl methanamine, 3,5-difluorobenzyl amine, 3,5-difluorophenyl methylamine, 3, 5-difluorobenzylamine, pubchem4409, 3,5-difluoro-benzylamine, 3,5-difluoro-benzyl amine
PubChem CID 145211
IUPAC Name 1-(3,5-difluorophenyl)methanamine
SMILES NCC1=CC(F)=CC(F)=C1

Specifications

Color Yellow
Boiling Point 184°C
Quantity 5 g
UN Number 2735
Formula Weight 143.14
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3,5-Difluorobenzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.