Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Diiodo-L-tyrosine Dihydrate 98.0+%, TCI America™
SDP

Catalog No. D061225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D061225G 25 g
D06125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D061225G Supplier TCI America Supplier No. D061225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18835-59-1
Molecular Formula C9H13I2NO5
Molecular Weight (g/mol) 469.01
MDL Number MFCD00150275
InChI Key YWAGQOOMOOUEGY-QVGJJETENA-N
Synonym 3,5-diiodo-l-tyrosine dihydrate, s-2-amino-3-4-hydroxy-3,5-diiodophenyl propanoic acid dihydrate, 3,5-diiodotyrosine dihydrate, 3,5-dijodtyrosindihydrat, c9h9i2no3.2h2o, 3,5-diiodo-l-tyrosine hydrate, l-tyrosine,3,5-diiodo-, hydrate 1:2, s-2-amino-3-4-hydroxy-3,5-diiodophenyl propanoicaciddihydrate, 2s-2-amino-3-4-hydroxy-3,5-diiodophenyl propanoic acid dihydrate, s-2-amino-3-4-hydroxy-3,5-diiodophenyl-propanoic acid dihydrate
PubChem CID 2724330
IUPAC Name (2S)-2-amino-3-(4-hydroxy-3,5-diiodophenyl)propanoic acid dihydrate
SMILES O.O.N[C@@H](CC1=CC(I)=C(O)C(I)=C1)C(O)=O

Specifications

Melting Point 213°C
Quantity 25 g
Formula Weight 432.98
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Diiodo-L-tyrosine Dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.